Learn More
Biomatik Corporation Dog Interferon Gamma (IFNg) ELISA Kit, 96 tests
Catalog No. 501502874
Encompass_Preferred
The Dog Interferon Gamma (IFNg) ELISA Kit allows for quantitative determination interferon Gamma in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P42161. Cross Activity: No significant cross-reactivity or interference between Interferon Gamma and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2874 Supplier Biomatik Corporation Supplier No. EKU05097
The Dog Interferon Gamma (IFNg) ELISA Kit allows for quantitative determination interferon Gamma in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P42161. Cross Activity: No significant cross-reactivity or interference between Interferon Gamma and analogues was observed. Restriction: For Research Use Only.