Learn More
Biomatik Corporation Human Integrin Alpha 11 (ITGa11) ELISA Kit
Supplier:  Biomatik Corporation EKU05028
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.125ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9UKX5. Target Synonyms: ITG-A11. Restriction: For Research Use Only.
Catalog No. 50-228-6104