Learn More
Biomatik Corporation Human Interferon Gamma (IFNg) ELISA Kit, 96 tests
Catalog No. 501502877
Encompass_Preferred
The Human Interferon Gamma (IFNg) ELISA Kit allows for quantitative determination interferon Gamma in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01579. Cross Activity: No significant cross-reactivity or interference between Interferon Gamma and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2877 Supplier Biomatik Corporation Supplier No. EKU05101
The Human Interferon Gamma (IFNg) ELISA Kit allows for quantitative determination interferon Gamma in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01579. Cross Activity: No significant cross-reactivity or interference between Interferon Gamma and analogues was observed. Restriction: For Research Use Only.