Learn More
Biomatik Corporation Human Integrin Alpha 5 (ITGa5) ELISA Kit
Supplier:  Biomatik Corporation EKU05034
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P08648. Target Synonyms: CD49e; CD49-E; ITG-A5; FNRA; VLA5A; Fibronectin Receptor,Alpha Polypeptide; Integrin alpha-F; CD49 antigen-like family member E. Restriction: For Research Use Only.
Catalog No. 50-228-6106