Learn More
Biomatik Corporation Pig Interferon Gamma (IFNg) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05103
Encompass_Preferred
Pig Interferon Gamma (IFNg) ELISA Kit for detecting IFNg in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 7.8-500pg/ml. Sensitivity: 2.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17803. Gene ID: 396991. Target Synonyms: IFN-G; IFG; IFI; INFr; IFN, Immune Interferon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2879