Learn More
Biomatik Corporation Human Interferon Gamma Induced Protein 10kDa (IP10) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05109
Encompass_Preferred
The Human Interferon Gamma Induced Protein 10kDa (IP10) ELISA Kit allows for quantitative determination interferon Gamma Induced Protein 10kDa in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.0 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02778. Cross Activity: No significant cross-reactivity or interference between Interferon Gamma Induced Protein 10kDa and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2883