Learn More
Biomatik Corporation Mouse Interferon Gamma Induced Protein 10kDa (IP10) ELISA Kit, 96 tests
Catalog No. 501502884
Encompass_Preferred
Mouse Interferon Gamma Induced Protein 10kDa (IP10) ELISA Kit for detecting IP10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q548V9. Gene ID: 15945. Target Synonyms: CXCL10; C7; IFI10; INP10; SCYB10; Crg2; GIP10; Mob1; Chemokine C-X-C Motif Ligand 10; Small Inducible Cytokine Subfamily B (Cys-X-Cys) Member 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2884 Supplier Biomatik Corporation Supplier No. EKU05110
Mouse Interferon Gamma Induced Protein 10kDa (IP10) ELISA Kit for detecting IP10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q548V9. Gene ID: 15945. Target Synonyms: CXCL10; C7; IFI10; INP10; SCYB10; Crg2; GIP10; Mob1; Chemokine C-X-C Motif Ligand 10; Small Inducible Cytokine Subfamily B (Cys-X-Cys) Member 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.