Learn More
Biomatik Corporation Rat Interferon Gamma Induced Protein 10kDa (IP10) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05111
Encompass_Preferred
Rat Interferon Gamma Induced Protein 10kDa (IP10) ELISA Kit for detecting IP10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48973. Gene ID: 245920. Target Synonyms: CXCL10; C7; IFI10; INP10; SCYB10; Crg2; GIP10; Mob1; Chemokine C-X-C Motif Ligand 10; Small Inducible Cytokine Subfamily B (Cys-X-Cys) Member 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2885