Learn More
Biomatik Corporation Human Interleukin 12 Receptor Beta 2 (IL12Rb2) ELISA Kit
Supplier:  Biomatik Corporation EKU05195
Encompass_Preferred
Human Interleukin 12 Receptor Beta 2 (IL12Rb2) ELISA Kit for detecting IL12Rb2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99665. Gene ID: 3595. Target Synonyms: IL12R-B2; IL12-RB2; RP11-102M16.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6127