Learn More
Biomatik Corporation Human Interferon Inducible T-Cell Alpha Chemoattractant (ITaC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05115
Encompass_Preferred
The Human Interferon Inducible T-Cell Alpha Chemoattractant (ITaC) ELISA Kit allows for quantitative determination interferon Inducible T-Cell Alpha Chemoattractant in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4000 pg/mL. Sensitivity: 26.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O14625. Cross Activity: No significant cross-reactivity or interference between Interferon Inducible T-Cell Alpha Chemoattractant and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2887