Learn More
Biomatik Corporation Human Interferon Regulatory Factor 3 (IRF3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05120
Encompass_Preferred
The Human Interferon Regulatory Factor 3 (IRF3) ELISA Kit allows for quantitative determination interferon Regulatory Factor 3 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.065 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q14653. Cross Activity: No significant cross-reactivity or interference between Interferon Regulatory Factor 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2888