Learn More
Biomatik Corporation Human Interleukin 18 Binding Protein (IL18BP) ELISA Kit
Supplier:  Biomatik Corporation EKU05258
Encompass_Preferred
Human Interleukin 18 Binding Protein (IL18BP) ELISA Kit for detecting IL18BP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.56ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95998. Gene ID: 10068. Target Synonyms: IL18BPa; MC51L-53L-54L Homolog Gene Product; Tadekinig-alfa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6132