Learn More
Biomatik Corporation Human Interferon Regulatory Factor 5 (IRF5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05123
Encompass_Preferred
The Human Interferon Regulatory Factor 5 (IRF5) ELISA Kit allows for quantitative determination interferon Regulatory Factor 5 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-10 ng/mL. Sensitivity: 0.235 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q13568. Cross Activity: No significant cross-reactivity or interference between Interferon Regulatory Factor 5 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2890