Learn More
Biomatik Corporation Mouse Interleukin 28A (IL28A) ELISA Kit
Supplier:  Biomatik Corporation EKU05310
Encompass_Preferred
Mouse Interleukin 28A (IL28A) ELISA Kit for detecting IL28A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q4VK74. Gene ID: 330496. Target Synonyms: IFNL2; Interferon,Lambda 2; Cytokine Zcyto20. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6140