Learn More
Biomatik Corporation Dog Interleukin 1 Alpha (IL1a) ELISA Kit, 96 tests
Catalog No. 501502891
Encompass_Preferred
The Dog Interleukin 1 Alpha (IL1a) ELISA Kit allows for quantitative determination interleukin 1 Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.0 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O46612. Cross Activity: No significant cross-reactivity or interference between Interleukin 1 Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2891 Supplier Biomatik Corporation Supplier No. EKU05127
The Dog Interleukin 1 Alpha (IL1a) ELISA Kit allows for quantitative determination interleukin 1 Alpha in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.0 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O46612. Cross Activity: No significant cross-reactivity or interference between Interleukin 1 Alpha and analogues was observed. Restriction: For Research Use Only.