Learn More
Biomatik Corporation Human Interleukin 29 (IL29) ELISA Kit
Supplier:  Biomatik Corporation EKU05312
Encompass_Preferred
Human Interleukin 29 (IL29) ELISA Kit for detecting IL29 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8IU54. Gene ID: 282618. Target Synonyms: IFNL1; IFN-λ1; Interferon Lambda 1; Cytokine Zcyto21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6142