Learn More
Biomatik Corporation Human Kininogen 1 (KNG1) ELISA Kit
Supplier:  Biomatik Corporation EKU05501
Encompass_Preferred
Human Kininogen 1 (KNG1) ELISA Kit for detecting KNG1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01042. Gene ID: 3827. Target Synonyms: BDK; KNG; HMWK; Alpha-2-Thiol Proteinase Inhibitor; Bradykinin; Kallidin; Fitzgerald factor; High molecular weight kininogen; Williams-Fitzgerald-Flaujeac factor; Ile-Ser-Bradykinin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6169