Learn More
Biomatik Corporation Human Interleukin 1 Beta (IL1b) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05140
Encompass_Preferred
The Human Interleukin 1 Beta (IL1b) ELISA Kit allows for quantitative determination interleukin 1 Beta in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01584. Cross Activity: No significant cross-reactivity or interference between Interleukin 1 Beta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2900