Learn More
Biomatik Corporation Rabbit Interleukin 1 Beta (IL1b) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05144
Encompass_Preferred
Rabbit Interleukin 1 Beta (IL1b) ELISA Kit for detecting IL1b in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14628. Gene ID: 100008990. Target Synonyms: IL1-B; IL1-Beta; IL1F2; IL-1β; Interleukin-1 Family Member 2; Catabolin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2904