Learn More
Biomatik Corporation Leukotriene B4 (LTB4) ELISA Kit
Supplier:  Biomatik Corporation EKU05632
Encompass_Preferred
Leukotriene B4 (LTB4) ELISA Kit for detecting LTB4 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 246.9-20,000pg/ml. Sensitivity: 90.1pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: LT-B4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6191