Learn More
Biomatik Corporation Mouse Interleukin 1 Family, Member 9 (IL1F9) ELISA Kit, 96 tests
Catalog No. 501502907
Encompass_Preferred
Mouse Interleukin 1 Family, Member 9 (IL1F9) ELISA Kit for detecting IL1F9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R460. Target Synonyms: IL-1F9; IL36g; IL-1H1; IL-1RP2; IL1E; IL1H1; IL1RP2; Interleukin 36, Gamma; Interleukin-1 Homolog 1; Interleukin-1 epsilon; Interleukin 1-Related Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2907 Supplier Biomatik Corporation Supplier No. EKU05149
Mouse Interleukin 1 Family, Member 9 (IL1F9) ELISA Kit for detecting IL1F9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R460. Target Synonyms: IL-1F9; IL36g; IL-1H1; IL-1RP2; IL1E; IL1H1; IL1RP2; Interleukin 36, Gamma; Interleukin-1 Homolog 1; Interleukin-1 epsilon; Interleukin 1-Related Protein 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.