Learn More
Biomatik Corporation Human Interleukin 1 Receptor Accessory Protein (IL1RAP) ELISA Kit, 96 tests
Catalog No. 501502908
Encompass_Preferred
The Human Interleukin 1 Receptor Accessory Protein (IL1RAP) ELISA Kit allows for quantitative determination interleukin 1 Receptor Accessory Protein in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.065 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NPH3. Cross Activity: No significant cross-reactivity or interference between Interleukin 1 Receptor Accessory Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2908 Supplier Biomatik Corporation Supplier No. EKU05150
The Human Interleukin 1 Receptor Accessory Protein (IL1RAP) ELISA Kit allows for quantitative determination interleukin 1 Receptor Accessory Protein in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.065 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NPH3. Cross Activity: No significant cross-reactivity or interference between Interleukin 1 Receptor Accessory Protein and analogues was observed. Restriction: For Research Use Only.