Learn More
Biomatik Corporation Dog Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit, 96 tests
Catalog No. 501502910
Encompass_Preferred
The Dog Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit allows for quantitative determination interleukin 1 Receptor Antagonist in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 31.25-2000 pg/mL. Sensitivity: 12.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9BEH0. Cross Activity: No significant cross-reactivity or interference between Interleukin 1 Receptor Antagonist and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2910 Supplier Biomatik Corporation Supplier No. EKU05154
The Dog Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit allows for quantitative determination interleukin 1 Receptor Antagonist in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 31.25-2000 pg/mL. Sensitivity: 12.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9BEH0. Cross Activity: No significant cross-reactivity or interference between Interleukin 1 Receptor Antagonist and analogues was observed. Restriction: For Research Use Only.