Learn More
Biomatik Corporation Human Interleukin 1 Receptor Like Protein 1 (IL1RL1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05165
Encompass_Preferred
The Human Interleukin 1 Receptor Like Protein 1 (IL1RL1) ELISA Kit allows for quantitative determination interleukin 1 Receptor Like Protein 1 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 62.5-4000 pg/mL. Sensitivity: 22.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q01638. Cross Activity: No significant cross-reactivity or interference between Interleukin 1 Receptor Like Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2912