Learn More
Biomatik Corporation Horse Interleukin 10 (IL10) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05175
Encompass_Preferred
The Horse Interleukin 10 (IL10) ELISA Kit allows for quantitative determination interleukin 10 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Equus caballus; Equine (Horse). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 6.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q28374. Cross Activity: No significant cross-reactivity or interference between Interleukin 10 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2917