Learn More
Biomatik Corporation Mouse Neuromedin B (NMB) ELISA Kit
Supplier:  Biomatik Corporation EKU06227
Encompass_Preferred
Mouse Neuromedin B (NMB) ELISA Kit for detecting NMB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.05pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9CR53. Gene ID: 68039. Target Synonyms: NM-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6257