Learn More
Biomatik Corporation Mouse Interleukin 10 (IL10) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05179
Encompass_Preferred
Mouse Interleukin 10 (IL10) ELISA Kit for detecting IL10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18893. Gene ID: 16153. Target Synonyms: CSIF; IL10A; TGIF; Cytokine Synthesis Inhibitory Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2920