Learn More
Biomatik Corporation Nicotinic Acid Adenine Dinucleotide Phosphate (NAADP) ELISA Kit
Supplier:  Biomatik Corporation EKU06298
Encompass_Preferred
Nicotinic Acid Adenine Dinucleotide Phosphate (NAADP) ELISA Kit for detecting NAADP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 123.5-10,000ng/ml. Sensitivity: 48.7ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6269