Learn More
Biomatik Corporation Human Nuclear Factor Of Activated T-Cells, Cytoplasmic 2 (NFATC2) ELISA Kit
Supplier:  Biomatik Corporation EKU06358
Encompass_Preferred
Human Nuclear Factor Of Activated T-Cells, Cytoplasmic 2 (NFATC2) ELISA Kit for detecting NFATC2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13469. Gene ID: 4773. Target Synonyms: NFAT1; NFATP; NF-ATP; NFAT pre-existing subunit; T-cell transcription factor NFAT1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6279