Learn More
Biomatik Corporation Rat Interleukin 11 (IL11) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05189
Encompass_Preferred
Rat Interleukin 11 (IL11) ELISA Kit for detecting IL11 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99MF5. Target Synonyms: AGIF; Adipogenesis Inhibitory Factor; Oprelvekin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2927