Learn More
Biomatik Corporation Human Pepsinogen A (PGA) ELISA Kit
Supplier:  Biomatik Corporation EKU06507
Encompass_Preferred
Sample Type: Serum, plasma, urine, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 3.12-200ng/mL. Sensitivity: 1.27ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P00790. Target Synonyms: PGA5; PG-A; Pepsinogen 5 Group I; Pepsinogen-5; Pepsin A-5. Restriction: For Research Use Only.
Catalog No. 50-228-6294