Learn More
Biomatik Corporation Gallus Interleukin 12B (IL12B) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05209
Encompass_Preferred
The Gallus Interleukin 12B (IL12B) ELISA Kit allows for quantitative determination interleukin 12B in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q6X0K9. Cross Activity: No significant cross-reactivity or interference between Interleukin 12B and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2932