Learn More
Biomatik Corporation Human Peptidylprolyl Isomerase C (PPIC) ELISA Kit
Supplier:  Biomatik Corporation EKU06526
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P45877. Target Synonyms: PPI-C; CYPC; Cyclophilin C; Peptidyl Prolyl Cis/Trans Isomerase C; Rotamase C. Restriction: For Research Use Only.
Catalog No. 50-228-6298