Learn More
Biomatik Corporation Human Phospholipase A2, Group X (PLA2G10) ELISA Kit
Supplier:  Biomatik Corporation EKU06621
Encompass_Preferred
Human Phospholipase A2, Group X (PLA2G10) ELISA Kit for detecting PLA2G10 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15496. Gene ID: 8399. Target Synonyms: GXPLA2; GX sPLA2; sPLA2-X; Group 10 secretory phospholipase A2; Phosphatidylcholine 2-acylhydrolase 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6310