Learn More
Biomatik Corporation Human Plasmin/Antiplasmin Complex (PAP) ELISA Kit
Supplier:  Biomatik Corporation EKU06654
Encompass_Preferred
Human Plasmin/Antiplasmin Complex (PAP) ELISA Kit for detecting PAP in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08697; P00747. Gene ID: 5345; 5340. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6313