Learn More
Biomatik Corporation Rabbit Platelet Factor 4 (PF4) ELISA Kit
Supplier:  Biomatik Corporation EKU06707
Encompass_Preferred
Rabbit Platelet Factor 4 (PF4) ELISA Kit for detecting PF4 in serum, platelet-poor plasma. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.19ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02776. Gene ID: 5196. Target Synonyms: CXCL4; SCYB4; Chemokine C-X-C-Motif Ligand 4; Oncostatin-A; Iroplact. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6322