Learn More
Biomatik Corporation Rat Interleukin 15 (IL15) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05228
Encompass_Preferred
Rat Interleukin 15 (IL15) ELISA Kit for detecting IL15 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97604. Gene ID: 25670. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2943