Learn More
Biomatik Corporation Human Progesterone Immunomodulatory Binding Factor 1 (PIBF1) ELISA Kit
Catalog No. 502286330
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.116ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8WXW3. Target Synonyms: PIBF; C13orf24; CEP90; Progesterone-Induced Blocking Factor 1. Restriction: For Research Use Only.
This item is not returnable. View return policy
Catalog No. 50-228-6330 Supplier Biomatik Corporation Supplier No. EKU06799
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.116ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8WXW3. Target Synonyms: PIBF; C13orf24; CEP90; Progesterone-Induced Blocking Factor 1. Restriction: For Research Use Only.