Learn More
Biomatik Corporation Dog Interleukin 17 (IL17) ELISA Kit, 96 tests
Catalog No. 501502946
Encompass_Preferred
The Dog Interleukin 17 (IL17) ELISA Kit allows for quantitative determination interleukin 17 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 7.81-500 pg/mL. Sensitivity: 2.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: C6L8D7. Cross Activity: No significant cross-reactivity or interference between Interleukin 17 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2946 Supplier Biomatik Corporation Supplier No. EKU05233
The Dog Interleukin 17 (IL17) ELISA Kit allows for quantitative determination interleukin 17 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 7.81-500 pg/mL. Sensitivity: 2.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: C6L8D7. Cross Activity: No significant cross-reactivity or interference between Interleukin 17 and analogues was observed. Restriction: For Research Use Only.