Learn More
Biomatik Corporation Human Proteasome Subunit Alpha Type 1 (PSMa1) ELISA Kit
Supplier:  Biomatik Corporation EKU06885
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P25786. Target Synonyms: HC2; NU; PROS30; Proteasome(Prosome,Macropain)Subunit,Alpha Type,1; Macropain subunit C2; Multicatalytic endopeptidase complex subunit C2; Proteasome nu chain. Restriction: For Research Use Only.
Catalog No. 50-228-6339