Learn More
Biomatik Corporation Rat Interleukin 17 (IL17) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05239
Encompass_Preferred
Rat Interleukin 17 (IL17) ELISA Kit for detecting IL17 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61453. Target Synonyms: IL17A; CTLA8; IL-17A; Cytotoxic T-Lymphocyte-Associated Protein 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2950