Learn More
Biomatik Corporation Human Interleukin 17C (IL17C) ELISA Kit, 96 tests
Catalog No. 501502954
Encompass_Preferred
The Human Interleukin 17C (IL17C) ELISA Kit allows for quantitative determination interleukin 17C in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9P0M4. Cross Activity: No significant cross-reactivity or interference between Interleukin 17C and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2954 Supplier Biomatik Corporation Supplier No. EKU05244
The Human Interleukin 17C (IL17C) ELISA Kit allows for quantitative determination interleukin 17C in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 7.8-500 pg/mL. Sensitivity: 3.3 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9P0M4. Cross Activity: No significant cross-reactivity or interference between Interleukin 17C and analogues was observed. Restriction: For Research Use Only.