Learn More
Biomatik Corporation Mouse Sodium/Glucose Cotransporter 1 (SGLT1) ELISA Kit
Supplier:  Biomatik Corporation EKU07385
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: SLC5A1; NAGT; Solute Carrier Family 5 Member 1; High affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 1. Restriction: For Research Use Only.
Catalog No. 50-228-6388