Learn More
Biomatik Corporation Mouse Interleukin 17C (IL17C) ELISA Kit, 96 tests
Catalog No. 501502955
Encompass_Preferred
Mouse Interleukin 17C (IL17C) ELISA Kit for detecting IL17C in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8K4C5. Gene ID: 234836. Target Synonyms: Cytokine CX2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2955 Supplier Biomatik Corporation Supplier No. EKU05245
Mouse Interleukin 17C (IL17C) ELISA Kit for detecting IL17C in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8K4C5. Gene ID: 234836. Target Synonyms: Cytokine CX2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.