Learn More
Biomatik Corporation Human Troponin T (TNT) ELISA Kit
Supplier:  Biomatik Corporation EKU07905
Encompass_Preferred
Human Troponin T (TNT) ELISA Kit for detecting TNT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13805; P4537; P45378. Gene ID: 7138; 7140. Target Synonyms: Tn-T; TNNT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6441