Learn More
Biomatik Corporation Rat Interleukin 17F (IL17F) ELISA Kit, 96 tests
Catalog No. 501502959
Encompass_Preferred
Rat Interleukin 17F (IL17F) ELISA Kit for detecting IL17F in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5BJ95. Gene ID: 301291. Target Synonyms: ML-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2959 Supplier Biomatik Corporation Supplier No. EKU05249
Rat Interleukin 17F (IL17F) ELISA Kit for detecting IL17F in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5BJ95. Gene ID: 301291. Target Synonyms: ML-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.