Learn More
Biomatik Corporation Dog Interleukin 18 (IL18) ELISA Kit, 96 tests
Catalog No. 501502960
Encompass_Preferred
The Dog Interleukin 18 (IL18) ELISA Kit allows for quantitative determination interleukin 18 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9XSR0. Cross Activity: No significant cross-reactivity or interference between Interleukin 18 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2960 Supplier Biomatik Corporation Supplier No. EKU05251
The Dog Interleukin 18 (IL18) ELISA Kit allows for quantitative determination interleukin 18 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 15.6-1,000 pg/mL. Sensitivity: 5.5 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9XSR0. Cross Activity: No significant cross-reactivity or interference between Interleukin 18 and analogues was observed. Restriction: For Research Use Only.