Learn More
Biomatik Corporation Human Interleukin 18 (IL18) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05253
Encompass_Preferred
The Human Interleukin 18 (IL18) ELISA Kit allows for quantitative determination interleukin 18 in Serum, Plasma, Urine, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q14116. Cross Activity: No significant cross-reactivity or interference between Interleukin 18 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2961