Learn More
Biomatik Corporation Mouse Interleukin 18 Binding Protein (IL18BP) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU05259
Encompass_Preferred
Mouse Interleukin 18 Binding Protein (IL18BP) ELISA Kit for detecting IL18BP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 24.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z0M9. Gene ID: 16068. Target Synonyms: IL18BPa; MC51L-53L-54L Homolog Gene Product; Tadekinig-alfa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2965