Learn More
Biomatik Corporation Uridine Triphosphate (UTP) ELISA Kit
Catalog No. 502286542
Encompass_Preferred
Uridine Triphosphate (UTP) ELISA Kit for detecting UTP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 61.7-5,000ng/ml. Sensitivity: 23.2ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
This item is not returnable. View return policy
Catalog No. 50-228-6542 Supplier Biomatik Corporation Supplier No. EKU08751
Add to Cart
Add to Cart
Uridine Triphosphate (UTP) ELISA Kit for detecting UTP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 61.7-5,000ng/ml. Sensitivity: 23.2ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.